missing translation for 'onlineSavingsMsg'
Learn More

Manganese(III) oxide, 98%, Thermo Scientific Chemicals

191.96 € - 673.35 €

Chemical Identifiers

CAS 1317-34-6
Molecular Formula Mn2O3
Molecular Weight (g/mol) 157.873
MDL Number MFCD00016217
InChI Key GEYXPJBPASPPLI-UHFFFAOYSA-N
Synonym manganese iii oxide, manganic oxide, dimanganese trioxide, manganese sesquioxide, manganese trioxide, manganese manganate, manganese sisquioxide, manganese 3+ oxide, manganese oxide, oxo oxomanganiooxy manganese
PubChem CID 14824
IUPAC Name oxo(oxomanganiooxy)manganese
SMILES O=[Mn]O[Mn]=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11325039
SDS View Documents
Thermo Scientific Alfa Aesar
087791.36
500 g
191.96 €
500g
Please to purchase this item. Need a web account? Register with us today!
11335039
SDS View Documents
Thermo Scientific Alfa Aesar
087791.A3
2 kg
673.35 €
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Manganese(III) oxide is used to produce ceramic magnets and semiconductors. Further, it serves as a precursor to lithium manganese oxide. In addition to this, it acts as a useful battery cathode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1317-34-6
157.873
GEYXPJBPASPPLI-UHFFFAOYSA-N
14824
O=[Mn]O[Mn]=O
Mn2O3
MFCD00016217
manganese iii oxide, manganic oxide, dimanganese trioxide, manganese sesquioxide, manganese trioxide, manganese manganate, manganese sisquioxide, manganese 3+ oxide, manganese oxide, oxo oxomanganiooxy manganese
oxo(oxomanganiooxy)manganese

Specifications

1080°C (decomposition)
Powder
Mn2O3
14,5737
Soluble in acid and ammonium chloride. Insoluble in water,alcohol and acetone.
O=[Mn]O[Mn]=O
157.873
157.86
4.50 g/mL
1317-34-6
98%
MFCD00016217
manganese iii oxide, manganic oxide, dimanganese trioxide, manganese sesquioxide, manganese trioxide, manganese manganate, manganese sisquioxide, manganese 3+ oxide, manganese oxide, oxo oxomanganiooxy manganese
GEYXPJBPASPPLI-UHFFFAOYSA-N
oxo(oxomanganiooxy)manganese
14824
98%
Manganese(III) oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332-H315-H319-H335

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P301+P312-P312-P405-P403+P233-P501a

Warning

missing translation for 'einecsNumber' : 215-264-4

missing translation for 'rtecsNumber' : OP0915000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.