missing translation for 'onlineSavingsMsg'
Learn More

Manganese(III) phosphate hydrate, 99%, Thermo Scientific Chemicals

Product Code. 11403478
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Insoluble in water.

Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104663-56-1
Molecular Formula MnO4P
Molecular Weight (g/mol) 149.91
MDL Number MFCD00209658
InChI Key BECVLEVEVXAFSH-UHFFFAOYSA-K
PubChem CID 71317435
SMILES [Mn+3].[O-]P([O-])([O-])=O

Specifications

Quantity 10 g
Solubility Information Insoluble in water.
IUPAC Name manganese(3+);phosphate;hydrate
Formula Weight 149.91 (Anhydrous)
Percent Purity 99%
Chemical Name or Material Manganese(III) phosphate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.