missing translation for 'onlineSavingsMsg'
Learn More

Manganese(IV) oxide, 80-85%, powder, Thermo Scientific Chemicals

25.80 € - 223.00 €

Chemical Identifiers

CAS 1313-13-9
Molecular Formula MnO2
Molecular Weight (g/mol) 86.94
MDL Number MFCD00003463
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
PubChem CID 14801
IUPAC Name dioxomanganese
SMILES O=[Mn]=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10185170
View Documents Promotion Details
Thermo Scientific Acros
213490250
25 g Plastic bottle
25.80 €
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10583571
View Documents Promotion Details
Thermo Scientific Acros
213490010
1 kg Plastic bottle
92.00 €
1kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10388703
View Documents Promotion Details
Thermo Scientific Acros
213490025
2.5 kg Plastic bottle
223.00 €
2.50kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

1313-13-9
86.94
NUJOXMJBOLGQSY-UHFFFAOYSA-N
14801
O=[Mn]=O
MnO2
MFCD00003463
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
dioxomanganese

Specifications

535.0°C
Gray
80-85%
MnO2
01,637; 02,257; 03,191; 04,317; 05,422; 06,357; 08,312; 13,233; 14,200; 15,197; 16,202
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
NUJOXMJBOLGQSY-UHFFFAOYSA-N
dioxomanganese
14801
80 to 85%
5.2g/mL
1313-13-9
Powder
MnO2
MFCD00003463
15, 5795
Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid
O=[Mn]=O
86.94
86.94
0.5% max. (105°C)
Manganese(IV) oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if inhaled.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
D

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 215-202-6

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only