Learn More
Manganese(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals
102.00 € - 291.00 €
Chemical Identifiers
CAS | 1313-13-9 |
---|---|
Molecular Formula | MnO2 |
Molecular Weight (g/mol) | 86.94 |
MDL Number | MFCD00003463 |
InChI Key | NUJOXMJBOLGQSY-UHFFFAOYSA-N |
Synonym | manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein |
PubChem CID | 14801 |
IUPAC Name | dioxomanganese |
SMILES | O=[Mn]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11339907
|
Thermo Scientific Alfa Aesar
042250.30 |
250 g |
102.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11349907
|
Thermo Scientific Alfa Aesar
042250.A1 |
1 kg |
291.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as an oxidizing agent in organic synthesis such as oxidation of allylic/benzylic alcohols, as a textile dye, as a reducing agent, and as a component of dry cell batteries such as zinc-carbon battery and alkaline battery. It is also used in making pigments for glasses and ceramics, and drier for paints. Further it is used in the manufacture of manganese steel and several manganese derivatives including potassium permanganate, a powerful versatile oxidant. Catalytic hydration of nitriles to amides by flowing through manganese dioxide has been reported. Manganese dioxide supported on inorganic oxide can be used for oxidation of methylamine through CWAO (Catalytic Wet Air Oxidation). It has high potential as highly efficient and robust material for water oxidation reaction (WORs).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1313-13-9 | |
86.94 | |
NUJOXMJBOLGQSY-UHFFFAOYSA-N | |
14801 | |
O=[Mn]=O |
MnO2 | |
MFCD00003463 | |
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein | |
dioxomanganese |
Specifications
535°C (decomposition) | |
Powder | |
MnO2 | |
14,5730 | |
Insoluble in water. | |
O=[Mn]=O | |
86.94 | |
86.94 | |
5.026 g/mL | |
Manganese(IV) oxide |
1313-13-9 | |
99.9% (metals basis) | |
MFCD00003463 | |
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein | |
NUJOXMJBOLGQSY-UHFFFAOYSA-N | |
dioxomanganese | |
14801 | |
99.9% | |
(metals basis) |
Safety and Handling
GHS H Statement
H302-H332
P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c
H302+H332-H373
missing translation for 'einecsNumber' : 215-202-6
missing translation for 'rtecsNumber' : OP0350000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Manganese(IV) oxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : manganese dioxide
RUO – Research Use Only