missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(IV) oxide activated, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
1 kg
Description
- Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%
- Variation in color and physical form is possible
Specifications
Specifications
Quantity | 1 kg |
UN Number | NONH for all modes of transport |
Formula Weight | 86.94g/mol |
Percent Purity | ≥90% |
Chemical Name or Material | Manganese(IV) oxide activated |
Safety and Handling
Product Identifier
- Manganese(IV) oxide activated
- Warning
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
- H302 + H332-Harmful if swallowed or if inhaled.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction