missing translation for 'onlineSavingsMsg'
Learn More

Manganese(IV) oxide activated, Honeywell Fluka™

Technical, activated,≥90% (RT)

58.26 € - 117.92 €

Chemical Identifiers

CAS 1313-13-9
Molecular Formula MnO2
Molecular Weight (g/mol) 86.94
MDL Number MFCD00003463
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
PubChem CID 14801
IUPAC Name dioxomanganese
SMILES O=[Mn]=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15686090
View Documents Promotion Details
Honeywell-Fluka
63548-250G
250 g
58.26 €
250g
Please to purchase this item. Need a web account? Register with us today!
15676090
View Documents Promotion Details
Honeywell-Fluka
63548-1KG
1 kg
117.92 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%
  • Variation in color and physical form is possible
Specifications

Chemical Identifiers

1313-13-9
86.94
NUJOXMJBOLGQSY-UHFFFAOYSA-N
14801
O=[Mn]=O
MnO2
MFCD00003463
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
dioxomanganese

Specifications

1313-13-9
MFCD00003463
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
O=[Mn]=O
86.94
86.94g/mol
Manganese(IV) oxide activated
MnO2
NONH for all modes of transport
NUJOXMJBOLGQSY-UHFFFAOYSA-N
dioxomanganese
14801
≥90%
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : OP0350000

Product Identifier
  • Manganese(IV) oxide activated

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H302 + H332-Harmful if swallowed or if inhaled.
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications