missing translation for 'onlineSavingsMsg'
Learn More

Manganese(IV) oxide activated, tech. 90%, Thermo Scientific Chemicals

Product Code. 15468979
Click to view available options
Quantity:
10 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Manganese(IV) oxide is suitable for use in batteries. It is widely used for the selective oxidation of allylic and benzylic alcohols to aldehydes or ketones, as a co-oxidant in combination with 1,4-benzoquinone and in the allylic oxidation of olefins catalyzed by Palladium(II)­ acetate. It acts a reagent for oxidations. It is the source of manganese and all its compounds, largely used in manufacturing of manganese steel, for making amethyst glass, decolorizing glass, painting on porcelain, faience and majolica. The precipitate is used in electrotechnics, pigments, browning gun barrels, drier for paints and varnishes, printing and dyeing textiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(IV) oxide is suitable for use in batteries. It is widely used for the selective oxidation of allylic and benzylic alcohols to aldehydes or ketones, as a co-oxidant in combination with 1,4-benzoquinone and in the allylic oxidation of olefins catalyzed by Palladium(II)­ acetate. It acts a reagent for oxidations. It is the source of manganese and all its compounds, largely used in manufacturing of manganese steel, for making amethyst glass, decolorizing glass, painting on porcelain, faience and majolica. The precipitate is used in electrotechnics, pigments, browning gun barrels, drier for paints and varnishes, printing and dyeing textiles.

Solubility
Insoluble in water.

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-13-9
Molecular Formula MnO2
Molecular Weight (g/mol) 86.94
MDL Number MFCD00003463
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
PubChem CID 14801
IUPAC Name dioxomanganese
SMILES O=[Mn]=O

Specifications

Melting Point ∽535°C (decomposition)
Quantity 10 g
Merck Index 14,5730
Solubility Information Insoluble in water.
Formula Weight 86.94
Percent Purity 90%
Grade Technical
Density 5.026
Chemical Name or Material Manganese(IV) oxide activated
compliance-icons compliance-icons
Product Identifier
  • Manganese (IV) oxide, activated
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Manganese oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.