missing translation for 'onlineSavingsMsg'
Learn More

Manganese(IV) oxide, Puratronic™, 99.996% (metals basis), Thermo Scientific Chemicals

67.44 € - 768.83 €

Chemical Identifiers

CAS 1313-13-9
Molecular Formula MnO2
Molecular Weight (g/mol) 86.94
MDL Number MFCD00003463
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
PubChem CID 14801
IUPAC Name dioxomanganese
SMILES O=[Mn]=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11326917
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010805.06
5 g
67.44 €
5g
Please to purchase this item. Need a web account? Register with us today!
11336917
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010805.14
25 g
242.21 €
25g
Please to purchase this item. Need a web account? Register with us today!
11346917
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010805.22
100 g
768.83 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as an oxidizing agent in organic synthesis such as oxidation of allylic/benzylic alcohols, as a textile dye, as a reducing agent, and as a component of dry cell batteries such as zinc-carbon battery and alkaline battery. It is also used in making pigments for glasses and ceramics, and drier for paints. Further it is used in the manufacture of manganese steel and several manganese derivatives including potassium permanganate, a powerful versatile oxidant. Catalytic hydration of nitriles to amides by flowing through manganese dioxide has been reported. Manganese dioxide supported on inorganic oxide can be used for oxidation of methylamine through CWAO (Catalytic Wet Air Oxidation). It has high potential as highly efficient and robust material for water oxidation reaction (WORs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1313-13-9
86.94
NUJOXMJBOLGQSY-UHFFFAOYSA-N
14801
O=[Mn]=O
MnO2
MFCD00003463
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
dioxomanganese

Specifications

535°C (decomposition)
Brown
99.997% (Metals basis)
MFCD00003463
Insoluble in water
O=[Mn]=O
86.94
86.94
5.026
1313-13-9
Powder
MnO2
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
NUJOXMJBOLGQSY-UHFFFAOYSA-N
dioxomanganese
14801
Odorless
Manganese(IV) oxide
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c

H302+H332-H373

missing translation for 'einecsNumber' : 215-202-6

missing translation for 'rtecsNumber' : OP0350000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only