missing translation for 'onlineSavingsMsg'
Learn More

Manganese phosphide, 99% (metals basis), Thermo Scientific Chemicals

35.48 € - 92.46 €

Chemical Identifiers

CAS 12263-33-1
Molecular Formula Mn3P2
Molecular Weight (g/mol) 226.76
MDL Number MFCD00064736
InChI Key ZZDYLDYDQHMVJZ-UHFFFAOYSA-N
IUPAC Name trimanganese(2+) diphosphanetriide
SMILES [P-3].[P-3].[Mn++].[Mn++].[Mn++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11351549
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014020.04
2 g
35.48 €
2g
Please to purchase this item. Need a web account? Register with us today!
11361549
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014020.09
10 g
92.46 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Manganese phosphide is used as a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12263-33-1
226.76
ZZDYLDYDQHMVJZ-UHFFFAOYSA-N
[P-3].[P-3].[Mn++].[Mn++].[Mn++]
Mn3P2
MFCD00064736
trimanganese(2+) diphosphanetriide

Specifications

1095°C
Powder
MFCD00064736
ZZDYLDYDQHMVJZ-UHFFFAOYSA-N
trimanganese(2+) diphosphanetriide
226.75
Odorless
Manganese phosphide
12263-33-1
Mn3P2
Insoluble in water.
[P-3].[P-3].[Mn++].[Mn++].[Mn++]
226.76
99%
(metals basis)
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only