missing translation for 'onlineSavingsMsg'
Learn More

Manganese pieces, irregular, 99.9% (metals basis), Thermo Scientific Chemicals

63.01 € - 376.88 €

Chemical Identifiers

CAS 7439-96-5
Molecular Formula Mn
Molecular Weight (g/mol) 54.94
MDL Number MFCD00011111
InChI Key PWHULOQIROXLJO-UHFFFAOYSA-N
Synonym colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
PubChem CID 23930
ChEBI CHEBI:35154
IUPAC Name manganese
SMILES [Mn]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11371298
SDS View Documents
Thermo Scientific Alfa Aesar
010236.30
250 g
63.01 €
250g
Please to purchase this item. Need a web account? Register with us today!
11351298
SDS View Documents
Thermo Scientific Alfa Aesar
010236.A1
1 kg
178.89 €
1kg
Please to purchase this item. Need a web account? Register with us today!
11361298
SDS View Documents
Thermo Scientific Alfa Aesar
010236.A7
5 kg
376.88 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7439-96-5
54.94
PWHULOQIROXLJO-UHFFFAOYSA-N
23930
manganese
Mn
MFCD00011111
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
CHEBI:35154
[Mn]

Specifications

Gray
Gray irregularly shaped pieces
99.9% (Metals basis)
MFCD00011111
PWHULOQIROXLJO-UHFFFAOYSA-N
manganese
23930
54.94
Manganese
7439-96-5
7.3g/cm3 at 20°C
Mn
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
[Mn]
54.94
CHEBI:35154
Broken Plate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-105-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.