missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese powder, APS <10 micron, 99.6% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
Color | Gray |
Appearance | Gray powder |
Assay Percent Range | 99.6% (Metals basis) |
Quantity | 50 g |
Form | Powder |
Chemical Name or Material | Manganese |
Sicherheit und Handhabung
Produktidentifikator
- Manganese powder
- Warning
- Flammable solid Category 2
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- MIXTURE LIST-Contains : Manganese
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'