missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese Standard, 1000mg/L in 2% HNO3 (Assurance Grade), SPEX CertiPrep™
33.15 € - 93.65 €
Chemical Identifiers
CAS | 7439-96-5, 7697-37-2, 7732-18-5 |
---|---|
Molecular Formula | Mn |
Molecular Weight (g/mol) | 54.94 |
MDL Number | MFCD00011111 |
InChI Key | PWHULOQIROXLJO-UHFFFAOYSA-N |
PubChem CID | 23930 |
ChEBI | CHEBI:35154 |
IUPAC Name | manganese |
SMILES | [Mn] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
16381236
|
SPEX Certiprep
PLMN2-2M |
30 mL | Triple-leached LDPE bottle |
33.15 €
30mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10060482
|
SPEX Certiprep
PLMN2-2T |
250 mL | Poly Bottle |
93.65 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Mn | 1000μg/mL |
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Chemical Identifiers
7439-96-5, 7697-37-2, 7732-18-5 | |
54.94 | |
PWHULOQIROXLJO-UHFFFAOYSA-N | |
CHEBI:35154 | |
[Mn] |
Mn | |
MFCD00011111 | |
23930 | |
manganese |
Specifications
7439-96-5, 7697-37-2, 7732-18-5 | |
0.1, 2.0, 97.9 | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00011111 | |
[Mn] | |
54.94 | |
CHEBI:35154 | |
2% HNO3 |
0.1, 2.0, 97.9 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Mn | |
PWHULOQIROXLJO-UHFFFAOYSA-N | |
manganese | |
23930 | |
Assurance | |
Manganese Standard |
Safety and Handling
Warning: Causes skin irritation, serious eye irritation Wear protective gloves/eye/face protection. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If skin/eye irritation occurs, get medical advice/attention.
missing translation for 'dotInformation' : Nonhazardous
Recommended Storage : Ambient
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 1000
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : nitric acid