missing translation for 'onlineSavingsMsg'
Learn More

Mannitol, Powder, Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 15538024
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
PubChem CID 6251
ChEBI CHEBI:16899
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

Packaging Poly Bottle
Quantity 2.5 kg
Specific Gravity 1L = 1.52kg
IUPAC Name hexane-1,2,3,4,5,6-hexol
Formula Weight 182.17
Grade Multi-Compendial/USP
Physical Form Powder
Chemical Name or Material Mannitol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'