Learn More
Thermo Scientific™ Mannitol Salt Agar
Presumptively detect, isolate and enumerate pathogenic staphylococci in milk, food, fish and other specimens while most other bacteria are inhibited.
17.50 €
Specifications
Description | Mannitol Salt Agar |
---|---|
Format | 90mm Monoplate |
Product Type | Agar |
Quantity | 10/Pk. |
Product Code | Brand | Format | Color | Content And Storage | Shelf Life | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Format | Color | Content And Storage | Shelf Life | Price | Quantity & Availability | |||
12929420
|
Thermo Scientific™
PO0151A |
90mm Monoplate | Red, clear | 2°C to 10°C | 9 Weeks |
17.50 €
Pack of 10 |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
Description
Presumptively detect, isolate and enumerate pathogenic staphylococci in milk, food, fish and other specimens while most other bacteria are inhibited, except a few halophilic species, with Thermo Scientific™ Mannitol Salt Agar. Look for colonies surrounded by bright yellow zones to recognize coagulase-positive staphylococci; non-pathogenic staphylococci produce colonies with reddish purple zones.
Clearly and easily differentiate Staphylococcus aureus and Staphylococcus epidermidis while most other bacteria is inhibited by the high salt concentration.Staphylococcus aureus is a coagulase-positive, catalase-positive, gram-positive cocci that occurs singly or in pairs, tetrads, short chains, and grape-like clusters. It is commonly carried on the skin and in the nose of healthy individuals but is also a major cause of infection in both healthcare and community settings worldwide. S. aureus can produce a variety of syndromes with clinical manifestations, including
skin and soft tissue infections, empyema, bloodstream infections, pneumonia, osteomyelitis, septic arthritis, endocarditis, sepsis, and meningitis.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Mannitol Salt Agar | |
90mm Monoplate | |
Agar | |
10/Pk. |
Safety and Handling
missing translation for 'shelfLife' : 9 Weeks
Your input is important to us. Please complete this form to provide feedback related to the content on this product.