Learn More
Hanna Instruments™ Manual Calibration Dissolved Oxygen Meter
Brand:  Hanna Instruments™ HI9142
Product Code. 10248660
  • 1180.00 € / Each
Explore more special offers
Specifications
HI76407 DO probe, HI76407A/P pre-formed PTFE DO membranes (2), HI7041S electrolyte solution (30 mL), batteries, and carrying case
0.1 mg/L
-5.0°C to +50.0°C
±0.2°C (±1°F) excluding probe error
300 g
2 years
Polarographic DO probe, internal temperature sensor, DIN connector
approx. 500 hours of use
0.0 to 19.9 mg/L (ppm)
±1.5% FS
0.1°C (1°F)
185 x 72 x 36 mm (7.3 x 2.8 x 1.4 in.)
10.6 oz.
1.5 V AA batteries (4)
Automatic, 0°C to 50°C (32°F to 122°F)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products