missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Manual Crimper, Steel, for Flip Top/Flip Off Seals or Flip Tear Up Seals

Crimping tools provide a reproducible, secure vial closure.

Brand:  Fisherbrand™ 28 06 0190

983.25 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11829440

  • 1045.00 € / Each
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Crimper
  • Manual
  • Steel
  • Non-autoclavable
  • Adjustable
TRUSTED_SUSTAINABILITY
Specifications

Specifications

28mm Crimper
Steel
For Flip Top/for Flip Off
28 mm
Not Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers