Learn More
Thermo Scientific™ Manual Crimpers and Decrimpers for Sealing and Unsealing Crimp Vials
Product Code. 17383763 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
11 mm Crimp Vials
13 mm Crimp Vials
20 mm Crimp Vials
8 mm Crimp Vials
Type:
Manual Crimper
Manual Decrimper
Unit Size:
Each
Product Code. 17383763
Brand: Thermo Scientific™ 60180DCR20
Use Thermo Scientific™ Manual Vial Crimpers and Decrimpers to attach or remove crimp caps from vials, respectively.
Use Thermo Scientific™ Vial Crimpers and Decrimpers to manually attach or remove crimp caps from vials, respectively. These manual crimpers and decappers are available in 8 mm, 11 mm, 13 mm, and 20 mm sizes and allow you to easily seal or remove crimp caps reliably.
TRUSTED_SUSTAINABILITY
Specifications
Type Manual Decrimper
For Use With (Equipment) 20 mm Crimp Vials
Material Stainless Steel/Aluminum
Product Line SureSTART Level 2