missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ MAP 2a,b (Microtubule-Associated Protein) Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ MAP 2a,b (Microtubule-Associated Protein) Ab-2, Mouse Monoclonal Antibody.

151.98 € - 570.00 €

Specifications

Antigen MAP 2a,b (Microtubule-Associated Protein) Ab-2
Clone AP20
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 4
Product Code Brand Form Quantity Price Quantity & Availability  
Product Code Brand Form Quantity Price Quantity & Availability  
12660697
View Documents
Epredia™
MS-249-S0
Concentrated 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12670697
View Documents
Epredia™
MS-249-S1
Concentrated 500 μL
446.00 €
500µL
Please to purchase this item. Need a web account? Register with us today!
12650697
View Documents
Epredia™
MS-249-S
Concentrated 1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12640697
View Documents
Epredia™
MS-249-R7
Prediluted 7 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Microtubule Associated Protein 2 exists in two high molecular weight forms (2a & 2b) and a low molecular weight form. The expression of MAP2 is developmentally regulated and its multiple forms arise by alternative splicing of a single gene. Ab-2 is highly specific to MAP2a,b and shows no cross-reaction with MAP1, MAP5, tubulin, or tau. Ab-2 reacts with dendrites and cell bodies of neurons but not with the neuronal processes. It does not react with the low molecular weight (70kDa) form (2c) of MAP2. It is useful in studies of neuron structure in normal and malignant brain tissue as well as in degenerative diseases such as Alzheimer's.

Host Species: Mouse

Clone: AP20

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: aa 997-1332

Immunogen: Bovine brain microtubule associated protein 2

Molecular Weight: ∽280kDa Doublet

Positive Control: Human glioblastoma T98G cells or brain

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

MAP 2a,b (Microtubule-Associated Protein) Ab-2
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RUO
Human
IgG1
AP20
Monoclonal
Mouse
Neuroscience
Bovine brain microtubule associated protein 2
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.