missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17490-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody can recognize both high MW and low MW isoforms of MAP2.
MAP2 (Microtubule Associated Protein 2) exists in two high molecular weight forms (MAP2a & MAP2b) and a low molecular weight form. The expression of MAP2 is developmentally regulated and its multiple forms arise by alternative splicing of a single gene. MAP2 is involved in microtubule assembly, which is an essential step in neurogenesis. The products of similar genes in rat and mouse are neuron-specific cytoskeletal proteins that are enriched in dendrites, implicating a role in determining and stabilizing dendritic shape during neuron development. MAP2 is useful in studies of neuron structure in normal and maligt brain tissue as well as in degenerative diseases such as Alzheimer′s Disease (AD).Specifications
MAP2 | |
Polyclonal | |
Unconjugated | |
MAP2 | |
MAP 2, MAP2, MAP2A, MAP2B, MAP2C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17756, 25595, 4133 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P11137, P15146, P20357 | |
Map2 | |
MAP2 Fusion Protein Ag11580 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction