missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MAP3K11 (MLK3), GST Tag Recombinant Protein
Human MAP3K11 (MLK3), GST Tag Recombinant Protein
634.00 € - 7890.00 €
Specifications
Accession Number | NP_002410.1 |
---|---|
Gene Alias | MAP3K11 (MLK3) |
KinaseFamily | MLK Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12634770
|
Invitrogen™
PR6962A |
100μg |
1826.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10342163
|
Invitrogen™
PV4009 |
1mg |
7890.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11593527
|
Invitrogen™
PV3788 |
10 μg |
634.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAP3K11 (MLK3) activates both the p38 MAP kinase and c-Jun kinase pathways.
Specifications
NP_002410.1 | |
MLK Kinase Family | |
Recombinant | |
Baculovirus |
MAP3K11 (MLK3) | |
Human | |
GST Tag |