missing translation for 'onlineSavingsMsg'
Learn More

Marimastat, Tocris Bioscience™

Product Code. 18015802
Click to view available options
Quantity:
1 mg
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Broad spectrum MMP inhibitor

Broad spectrum inhibitor of MMPs (IC50 values are 3, 5, 6, 9 and 13 nM for MMP-9, MMP-1, MMP-2, MMP-14 and MMP-7 respectively). Inhibits peritoneal dissemination of human gastric cancer cells in vivo through inhibition of tumor angiogenesis.

Chemical Identifiers

CAS 154039-60-8
Molecular Formula C15H29N3O5
Molecular Weight (g/mol) 331.413
InChI Key OCSMOTCMPXTDND-OUAUKWLOSA-N
Synonym marimastat, marimastat usan, marimastat usan/inn, unii-d5eqv23tds, d5eqv23tds, 2s,3r-3-1s-2,2-dimethyl-1-methylcarbamoxy propyl carboyl-2-hydroxy-5-methylhexanohydroxamic acid, 2s,3r-3-1s-2,2-dimethyl-1-methylcarbamoyl propyl carbamoyl-2-hydroxy-5-methylhexanohydroxamic acid, 2s,3r-n 4-2s-3,3-dimethyl-1-methylamino-1-oxobutan-2-yl-n 1 ,2-dihydroxy-3-2-methylpropyl butanediamide, 2r,3s-n-2s-3,3-dimethyl-1-methylamino-1-oxobutan-2-yl-n',3-dihydroxy-2-2-methylpropyl butanediamide, 2s,3r-n-1s-2,2-dimethyl-1-methylcarbamoyl propyl-n',2-dihydroxy-3-2-methylpropyl butanediamide
PubChem CID 119031
ChEBI CHEBI:50662
IUPAC Name (2R,3S)-N-[(2S)-3,3-dimethyl-1-(methylamino)-1-oxobutan-2-yl]-N',3-dihydroxy-2-(2-methylpropyl)butanediamide
SMILES CC(C)CC(C(C(=O)NO)O)C(=O)NC(C(=O)NC)C(C)(C)C

Spezifikation

Quantity 1 mg
Formula Weight 331.41
Percent Purity >99%
Chemical Name or Material Marimastat
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'