missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MarrowMAX™ Bone Marrow Medium
GREENER_CHOICE

Fully-supplemented MarrowMAX™ Bone Marrow Medium for short-term culture of bone marrow and other hematopoietic cells for cytogenetic studies and in vitro diagnostic procedures.

Brand:  Gibco™ 12260001

 View more versions of this product

Product Code. 11380882

  • 1158.00 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Gibco MarrowMAX Bone Marrow Medium is a fully-supplemented medium developed for the short term culture of bone marrow and other hematopoietic cells for cytogenetic studies and in vitro diagnostic procedures.

MarrowMAX Bone Marrow Medium features:

  • Superior performance
  • Ready-to-use format
  • Quality and performance testing
  • Unique, optimized formulation

Superior performance

MarrowMAX Bone Marrow Medium outperforms other commercially available media containing Giant Cell Tumor (GCT) conditioned media, providing consistent performance with a higher mitotic index and superior chromosomal morphology.

Ready-to-use format

MarrowMAX Bone Marrow Medium is a frozen, 1X medium, ready to use upon thawing. No supplementation is required. Thawed medium can be stored at 2–8°C for up to 60 days.

Quality and performance testing

Every lot of MarrowMAX Bone Marrow Medium is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance. Bone marrow cells from the aspirate of a normal adult donor are cultured for 24 hours in MarrowMAX Bone Marrow Medium before measuring the mitotic index and chromosome banding resolution. In addition, each lot must pass a sterility test prior to release.

Unique, optimized formulation

MarrowMAX Bone Marrow Medium is an optimized RPMI 1640 medium, supplemented with Fetal Bovine Serum (FBS), gentamicin, and L-glutamine. It also contains a novel human stromal cell conditioned medium that consists of a unique blend of hematopoietic growth factors. MarrowMAX Bone Marrow Medium does not contain GCT conditioned medium.

cGMP Manufacturing and Quality System

MarrowMAX Bone Marrow Medium is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: -5°C to -20°C. Protect from light.
Shipping conditions: Frozen
Shelf life: 18 months from date of manufacture. Thawed medium can be stored at 2°C to 8°C for up to 60 days.
1 X
Bone Marrow Medium
Sterile-filtered
Gentamicin
100 mL
Human
Bone Marrow, Hematopoietic Cell
Liquid
Standard Serum Supplementation
Glutamine, Phenol Red, FBS, Conditioned Medium (from Stromal Cells)
MarrowMAX™
Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.