Learn More
Masterflex™ Mass Flowmeter
Economical, highly accurate, and rapid flowmeter with a miniature coil design. Cole-Parmer™ Mass Flowmeter provides precise and repeatable measurements.
Brand:  Masterflex™ 32707-32
Product Code. 15128016
  • 1075.00 € / Each
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Thermal gas flow sensing technique
  • Calibrated to air/nitrogen
  • Cable hub connector for a quick connection to a power supply and 0 to 5 VDC output cable; order power supplies/cables under separately
  • Low pressure drop across the sensor
  • Output signal can connect to a remote display, recorder, or any instrument that accepts a 0 to 5 VDC signal
  • Compact size makes it easy to carry
  • Wetted materials are anodized aluminum, 316 stainless steel, Viton™ O-rings, and acetal fittings.
Specifications
Specifications
Mass Flowmeter
Air, Nitrogen
1/4 in. Compression Fittings
2 LPM
10°C to 50°C
±1.5% (Full Scale)
150 psig
3.5-digit LCD
±0.5% (Full Scale)