Learn More
Atago™ MASTER-BC Model Battery Coolant Checker Refractometer
Use this refractometer for a variety of applications. Atago™ MASTER-BC Model Battery Coolant Checker Refractometer has an exclusive scale for measuring specific gravity of battery fluid and for checking the freezing temperature of ethylene glycol and propylene glycol.
Brand:  Atago™ 2931
Product Code. 11496168
  • 279.00 € / Each
Explore more special offers
Description
Description
  • For use with chemicals found in antifreeze mixtures in automobiles, thermal catalysts for solar power systems and other industrial applications
  • MASTER-BC is the successor model to the conventional BC-2E model
  • Scale is graduated in °C
  • Constructed of durable material suitable for salty and acid samples
Specifications
Specifications
Battery Coolant Refractometer
MASTER-BC
Measuring specific gravity of battery fluid and checking the freezing temperature of ethylene glycol and propylene glycol which are used as antifreeze mixture in automobiles
0 to -50°C Ethylene Glycol/Propylene Glycol
1.150 to 1.300 Battery Fluid d20/20
Freezing temperature, Battery fluid d20/20
90 g