Learn More
Atago™ MASTER-S10M Model Salinity Refractometer
Designed to control the concentration of saltwater, such as in cooking or rinsing seafood. Atago™ MASTER-S10M Model Salinity Refractometer utilizes a new material that is durable against salty and acidic samples.
Brand: Atago™ 2473
This item is not returnable.
View return policy
Description
- For use when water resistance is not a priority and the temperature compensation will be carried out manually
- Easy to read and hold
- Designed for smooth sampling
- Easy disposal of sample from the edge
- Improved grip for better hygiene
- Balanced structure
- Streamlined body and the strong grip with rich texture
Specifications
Salinity Refractometer | |
MASTER-S10M | |
Designed to control the concentration of saltwater, such as in cooking or rinsing seafood, and utilizes a new material that is durable against salty and acidic samples |
0 to 10 g/100 g Sodium Chloride | |
Sodium chloride | |
105 g |