missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ C-Flex™ I/P™ High-Performance Precision Pump Tubing

Ensure optimal Masterflex™ pump performance with this tubing. Masterflex™ C-Flex™ I/P™ High Performance Precision Pump Tubing combines biocompatibility of silicone with chemical resistance similar to Tygon™. Ideal for applications involving pressure, suction lift and viscous fluids.

Brand:  Masterflex™ 06424-70

159.16 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11761209

  • 164.00 € / Each
Estimated Shipment: 26-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Exceptional tensile and tear strength, good biocompatibility, low gas permeability and a smooth surface
  • General purpose lab tubing for applications requiring high performance and long life
  • Thicker wall compared to Precision pump tubing
  • Lot-to-lot consistency for superior accuracy and repeatability
  • Provides longer life than other platinum and peroxide silicone tubing formulations
  • Very low protein binding
  • Heat sealable, weldable
  • Better resistance to pH fluctuation
  • Complies with USP Class VI, EP, REACH, RoHS, ADCF
  • Economical
  • Opaque White
Specifications

Specifications

Autoclavable
C-Flex
For general-purpose lab uses requiring high performance and long life
70
3/8 in.
Low
40 psi
135°C
9.5 mm
760 cm
-40°C
0.01 to 8L/min.
8.8 m
Opaque White
660 mm
Pump Tubing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.