Learn More
Masterflex™ I/P™ Precision Modular Pump
Purge tubing before or after pumping and pump fluid in either direction. Masterflex™ I/P™ Precision Modular Pumps allow you the flexibility to separate the motor from the controller. Designed to fit in limited or awkward spaces.
Brand:  Masterflex™ 77962-30
6205.29 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 13487099
  • 6370.00 € / Each
Explore more special offers
Description
Description
  • Ideal for metering and critical transfer applications
  • These accurate modular drives have powerful motors, speed display and ±0.25% speed control for precise flow regulation
  • Splash-resistant ABS plastic controller housing is IP33 rated; drive housing is epoxy finished steel, IP33 rated
  • Powerful motors let you stack two pump heads for increased flow rates and for multichannel applications
  • Remote control capabilities via DB9 female connector on back of drive
  • Speed control input 4 to 20mA or 0 to 10V
  • Pump direction (CW/CCW) via open collector or contact closure; Start/stop via open collector or contact closure
  • Modular drive lets you place motor and controller where convenient
  • Three-digit LED display shows motor rpm
  • Membrane keypad for easy up-front control of speed, direction, and start/stop
  • Reversible motor
  • LED indicators confirm motor direction
Specifications
Specifications
CE
LED
8 L/min.
Yes
Aluminum (Controller), Aluminum (Drive), Steel (Drive)
Precision washdown modular drive with wall-mount controller
± 0.25%
361.95 mm
177.8 mm
165.1 mm
90/260 VAC
50/60 Hz
Variable Speed
IP66
650 rpm
07591-30 Modular Driver, 6 ft. (1.8 m) Line Cord with IEC 320/CEE22 Socket, C-Flex ULTRA I/P 73 06434-73 10 ft. (3 m) Tubing, Easy-Load 77602-10 Pump Head
I/P Easy-Load
I/P Easy-Load pump head 77602-10
17.25 in.
7 in.
6.5 in.
4.4 A 115/230 VAC 50/60 Hz
4.4 A