missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ I/P™ Series Peristaltic Pump

A complete digital console process pump system with Easy-Load™ pump head for precise fluid transfer and dispensing. The I/P™ Series Peristaltic Pump allows for quick and effortless tubing changes without the need for tools or complex procedures.

Brand:  Masterflex™ 77964-20

9200.64 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 14803762

  • 9240.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Three dispensing modes
  • Maintenance free, easy to clean
  • Graphical LCD display shows four operating modes - continuous run, timed dispense, copy dispense, volume dispense; view pump performance continually
  • Menu-driven interface lets you set one of seven langauges for programming and operation
  • Dispense by mL
  • Anti-drip function ensures dispense accuracy
  • Reversible motor - purge tubing before or after pumping; pump fluid in either direction
  • Brushless motor offers ±0.1%speed control accuracy with a 6000 to 1 turndown ratio
  • Powerful motors let you stack two pump heads with any tubing formulation for increased flow rates and for multichannel applications
  • Remote control capabilities via watertight connection
Specifications

Specifications

CE, cETLus, ISO 1900:2008
Variable Speed
IP66
650 rpm
3/8 hp
Stainless Steel (Case)
500.13 mm
282.7 mm
327.15 mm
C-Flex ULTRA
LCD
8 L/min.
Yes
3 m (10 ft.) Tubing, 77420-20 Drive, C-Flex ULTRA I/P 73 06434-73, I/P Easy-Load 77601-10 Pump Head
19.7 in.
11.1 in.
12.9 in.
115/230 VAC
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.