missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ L/S™ Analog Variable-Speed Console Drive Systems

Compact, cost effective pump systems for general fluid transfer applications.

Brand:  Masterflex™ 77910-75

 View more versions of this product

Product Code. 16408675

  • 1750.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Reversible motor lets you easily prime/purge tubing and pump in either direction
  • Remote control of start/stop via DB9 connector on back of drive
Specifications

Specifications

1
IP22
20 to 600 rpm
07555-05 (L/S™ Console Drive), 07514-12 (L/S™ Easy-Load Pump Head), 06434-24, 10 ft. (C-FLEX™ Ultra, L/S™ 24)
Easy Load
230 VAC
Analog
56 to 1700 mL/min.
Start/Stop
Painted steel and ABS plastic
29.2 x 18.4 x 13.34 cm
0.8 A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.