missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ L/S™ Digital Process Drives

Transfer, dispense, and meter in lab process and washdown environments.

Brand:  Masterflex™ 07575-50

 View more versions of this product

Product Code. 16448665

  • 6970.00 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Flow rate 0.006 to 3400 mL/min with L/S tubing—flow rate depends on drive rpm pump head, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating mode— continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication—voltage, current and contact closure
  • Dispense by Volume in mL (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)
  • Antidrip function ensures dispensing accuracy
  • Models with open-head sensor stops the drive when the pump head is opened (when used with sensor-equipped pump head)
  • NEMA 4X (IP66) rated housing for washdown applications
Specifications

Specifications

±0.1% (0.01 rpm at 100 rpm)
Connection Types: Ethernet/IP, Bluetooth LE, USB
90 to 260 VAC
6 ft. (1.8 m) line cord with IEC 320/CEE 22 connector coupler; watertight cord connection. Specify destination country when ordering; drive is shipped with country-specific plug/cord set.
0.1 to 600 rpm
2.2 A
LCD
0.006 to 3400 mL/min.
Non-sterile
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers