Learn More
Masterflex™ Multi-Channel Large Cartridge Pump Head
Use the cartridges for synchronous, multichannel pumping. MasterFlex™ L/S™ Large Cartridges feature accurate flow rates and adjustable tubing retainers to hold tubing in place.
Brand:  Masterflex™ 07519-70
 View more versions of this product
Product Code. 11868443
  • 142.00 € / Each
Estimated Shipment: 28-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed for use with Masterflex pump head models: 07519-05 and -06
  • Set tubing retainers to your tube size and you are ready to pump
  • On small cartridges, optimum retention is preset; simply load tubing and start pumping
  • Each cartridge accepts microbore and/or L/S precision pump tubing, please refer to table below
  • All cartridges have adjustable occlusion which maintains a bi-directional flow variation of 5%
  • Tighten occlusion for higher pressures, loosen for longer tubing life
  • Combine the input/output of two cartridges to reduce pulsation
  • Cartridges are constructed from polycarbonate and glass-filled nylon
Mix small and large cartridges on pump heads that accept both sizes; two small cartridges occupy the same space as one large cartridge
Only Silicone and C-Flex tubing formulations are recommended for size L/S 17 tubing when used in L/S cartridge pump heads.
Specifications
Specifications
Microbore and Precision Tubing
Adjustable
Cartridge Pump Head
Multiple
1 Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Masterflex™ Multi-Channel Large Cartridge Pump Head > For use with: L/S 07519-05, 07519-06 Cartridge Pump Heads, For Synchronous, Multi-channel Pumping