Learn More
Masterflex™ L/S™ Compact Variable-Speed Pump Drive
Use these drives when you need to pump fluid at a higher rate, but space is limited in your lab. MasterFlex™ L/S™ Compact Variable-Speed Pump Drive delivers versatile performance in limited space.
Brand:  Masterflex™ 77200-20
 View more versions of this product
Product Code. 11791349
  • 1000.00 € / Each
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Drive accepts two different pump head types: Standard and Easy-Load™. Separate single-turn speed control and on/off switch with green power indicator maintain speed setting while turning pump on/off. Stackable, compact drive is perfect for small work areas.
  • Painted steel housing is IP22 rated. Housing protects drive from vertically falling water
  • MAX button runs pump at maximum rpm—use to prime or to rapidly flush tubing
  • Reversible permanent magnet DC motor lets you pump in either direction
  • Remote control options on the back of pump
  • Start/stop pump via contact closure
Specifications
Specifications
115/230 VAC 120 mA
114 mm
132 mm
±5%
115/230 VAC
2.1 to 560 mL/min.
13.6 cm
Yes
Pump Drive
35–200 rpm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products