missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ PharMed™ BPT I/P™ Precision Pump Tubing
Use this pump tubing for cell and tissue culture work and pharmaceutical applications. Masterflex™ PharMed™ BPT I/P™ Precision Pump Tubing resists acids, alkalies, oxidizing agents, and animal and vegetable oils. Tubing is heat-sealable, bondable and formable. Good for light-sensitive samples.
Brand: Masterflex™ 06508-82
Description
Use this pump tubing for cell and tissue culture work and pharmaceutical applications. Masterflex™ PharMed™ BPT I/P™ Precision Pump Tubing resists acids, alkalies, oxidizing agents, and animal and vegetable oils. Tubing is heat-sealable, bondable and formable. Good for light-sensitive samples.
- Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory Approvals to match your unique application
- Always in stock — most formulations and sizes available to ship same day
- Economical tubing with a longer tubing life
Specifications
Pump Tubing | |
0.5 in. | |
PharMed BPT | |
82 mm | |
760 cm | |
-51°C | |
Acids, Alkalies | |
0.02 to 13 mL/min. | |
Opaque Beige | |
660 mm Hg |
Autoclavable | |
12.7 mm | |
3.22 in. | |
24.9 ft. | |
For cell and tissue culture work and pharmaceutical uses. Also good for light-sensitive samples. | |
1 Ea. | |
82 | |
1/2 in. | |
20 psi | |
132°C |