Learn More
Masterflex™ Platinum-Cured Silicone Tubing
Ensure optimal performance from your Masterflex™ pump. Masterflex™ Platinum-Cured Silicone Tubing is manufactured to extremely close tolerances and ideal for pharmaceutical and biotechnological use.
Brand:  Masterflex™ MF0316
Product Code. 10128791
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Slightly greater clarity than peroxide-cured tubing.
  • Smooth surfaces; lower protein binding levels.
  • Fewer potential leachables.
  • Ideal for pharmaceutical and biotechnology use.
Specifications
Specifications
9.5 mm
-50°C
8000 mL/min maximum
40psi, 2.7 bar
230°C
Silicone (Platinum-Cured)
73
7.6 m
660 mm Hg maximum