Learn More
Masterflex™ L/S™ Platinum-Cured Silicone Pump Tubing, spooled
Reduce scrap with spooled tubing. Masterflex™ L/S™ Platinum-Cured Silicone Pump Tubing features a smooth surface and low protein binding levels, ideal for pharmaceutical, biotechnology, peristaltic pumping and food applications.
Brand:  Masterflex™ 96403-13
 View more versions of this product
Product Code. 11715158
  • 1370.00 € / Each
Currently Unavailable: We have no information when this product will be available
View alternative products
Explore more special offers
Description
Description
Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.
  • Ensure that you always have enough tubing on hand
  • Slightly greater clarity
  • Preferred by pharmaceutical and biotechnology industries
  • Fewer potential leachables
  • USP Class VI
  • FDA compliant
Specifications
Specifications
FDA approved; Prop 65 NA (no Prop 65 chemicals)
0.8 mm
500 ft.
-40°C
36 mL/min.
Gamma Irradiation
660 mm Hg
Pump Tubing
Platinum-cured Silicone
152.4 m
13
1/16 in.
40 psi, 2.7 bar
100°C