Learn More
Masterflex™ Tygon™ E-LFL Pump Tubing
Longest life of all Tygon™ formulations
Brand: Masterflex™ 06449-34
This item is not returnable.
View return policy
Description
- Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Regulatory approvals to match your unique application
Specifications
Pump Tubing | |
1.42 mm | |
3000 cm | |
-46°C | |
34 |
Autoclavable | |
Tygon E-LFL | |
general laboratory use and provides longer life with peristaltic tubing pumps | |
1 Each | |
74°C |