Learn More
Materials Kit
Brand:  Edulab MATKIT
Product Code. 13132800
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
2 each of seventeen blocks: 50 x 40 x 30 mm: softwood, hardwood, paraffin wax, aluminum, iron and foam polystyrene; 20 x 20 x 100 mm: perspex, glass, slate, aluminum, softwood, marble; 50 x 50 x 20 mm: lead; 50 x 50 x 80 mm: aluminum; 50 x 50 x 20
Familiarize students with appearance, feel, texture, hardness and density of a range of common substances