missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Matrigel™ Growth Factor Reduced (GFR) Basement Membrane Matrix, Phenol Red-Free, LDEV-Free

Solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma. Corning™ Matrigel™ Growth Factor Reduced (GFR) Basement Membrane Matrix, Phenol Red-Free, LDEV-Free is tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

2344.30 € - 5180.00 €

Specifications

Content And Storage Store at -20°C. Avoid multiple freeze-thaws. Do not store in frost-free freezer. Keep frozen.
Without Additives No Phenol Red, No LDEV
For Use With (Application) Cell Growth and Differentiation, Metabolism/Toxicology Studies, Invasion Assays, In Vitro and In Vivo Angiogenesis Assays, In Vivo Angiogenesis Studies and Augmentation of Tumors in immunosuppressed Mice
Source Engelbreth-Holm-Swarm (EHS) Mouse Tumor
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15585729
View Documents
Corning™
356238
50 mL (10 mL x 5) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15595729
View Documents
Corning™
356239
100 mL (10 mL x 10) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

Specifications

Specifications

Store at -20°C. Avoid multiple freeze-thaws. Do not store in frost-free freezer. Keep frozen.
No Phenol Red, No LDEV
Cell Growth and Differentiation, Metabolism/Toxicology Studies, Invasion Assays, In Vitro and In Vivo Angiogenesis Assays, In Vivo Angiogenesis Studies and Augmentation of Tumors in immunosuppressed Mice
Engelbreth-Holm-Swarm (EHS) Mouse Tumor
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.