Learn More
Corning™ Matrigel™ Matrix for Organoid Culture
BIOPHARMAGREENER_CHOICE
Corning™ Matrigel™ Matrix for Organoid Culture is a solubilized basement membrane preparation that has been optimized for organoid culture.
Brand:  Corning™ 356255
679.25 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Product Code. 16491434
  • 715.00 € / 10mL
Explore more special offers
Description
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
-20°C
Stem Cell Media
10 mL
Lot dependent
Organoid Culture and workflow
Engelbreth-Holm-Swarm (EHS) mouse tumor