missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Matrix™ 2D Barcoded Glass Storage Tubes, Matrix 1.0 mL, 2D barcoded, glass, 5 Latch Racks of 96 tubes/case
GREENER_CHOICE

Guarantee reliability in lab applications with these 2D barcoded glass tubes in a safe ANSI-footprint latch rack.

Brand:  Thermo Scientific™ 3850

820.80 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.


Product Code. 11839130

  • 1140.00 € / Pack of 480
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Store and track robust sample volumes with Thermo Scientific™ Matrix™ 2D Barcoded Glass Storage Tubes. The proprietary barcoding process produces a permanent, high contrast 2D barcode, offering maximum traceability of compound, biological and genomic samples. Available with a range of sealing options and used with Thermo Scientific™ VisionMate™ and labeling systems, these barcoded storage tubes can improve a range of sample management needs and workflows.

Secure Tracking
  • A permanently-bonded,unique 2D barcode is laser-etched onto the base of every Matrix 2D barcoded storage tube to securely identify and track samples
  • Corresponding with a laboratory's throughput needs, VisionMate 2D barcode readers instantly decode each tube's 2D barcode
  • A range of VisionMate 2D barcode readers are available to meet your application and throughput needs
  • Scan the tube into any application or database, or integrate via various methods—the choice is yours
  • VisionMate systems are flexible enough to allow 2D barcode data to be integrated into various applications, databases, and automated systems
  • Once sample data is stored, just scan the tube using the database
  • Find feature to instantly access sample information and identify the contents of each storage tube

Superior Storage Rack Design

  • Unlike traditional storage tubes or blocks, Matrix 96-format 2D barcoded storage tubes are available in a proprietary specially-designed, stackable, microplate footprint
  • Latch Racks to save precious space in your freezer, refrigerator, compound store or on the bench top
  • The latch rack design enables manual multichannel pipette access to 2D tubes and eliminates contamination risk with lid design that does not contact the benchtop
  • The latch rack lid can be lifted robotically for 2D tube access with automated liquid handling systems and high throughput applications

Stringent Quality Control

  • Every 2D barcoded storage tube is scanned to guarantee readability and each code is checked against a full database of all previously assigned 2D codes to ensure zero duplicates across the entire line of Matrix 2D barcoded storage tubes
  • Every storage tube is leak tested to ensure the integrity and security of samples

Storage Tracking Solutions for Any Laboratory

  • Glass tubes can be used for storing samples in DMSO, chlorinated solvents and other harsh chemicals and applications where newly synthesized compounds that need to be dried down or lyophylized
  • Custom 2D coding options are available to meet your laboratory's individual database or tracking requirements
  • 2D storage tubes are compatible with Thermo Scientific freezer racks that allow optimal utilization of storage space

Recommended for:

  • Individual sample retrieval
  • Instant sample identification
  • Secure identification and shipping of archived samples
  • Storage of samples
  • Tracking of samples
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2D barcoded
Glass
2D Barcode
5 Case (Latch Racks of 96 Tubes)
1 mL
5 Latch Racks of 96 tubes/case
Matrix™
0.0338 fl oz.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.