missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Matrix™ 2D Barcoded Low Volume ScrewTop Storage Tubes
GREENER_CHOICE

Maximize storage capacity with these space-saving, 2D barcoded, low volume, screwtop storage tubes, the solution for secure sample storage.

Brand:  Thermo Scientific™ 3747

340.99 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15432322

  • 396.00 € / Pack of 480
Estimated Shipment: 03-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Maximize storage capacity with the space-saving Thermo Scientific™ Matrix 2D Barcoded Low Volume ScrewTop Storage Tubes, now available in a barcoded rack. These 200μL 2D internally-threaded tubes maximize capacity in manual or automated storage systems. They can be used for storage in virtually any laboratory condition, from room temperature through the vapor phase of liquid nitrogen. Permanent 2D barcodes enable safe, secure tracking of valuable small-volume samples. Compatible with the complete line of Thermo Scientific 2D barcode readers, sample data can be integrated into existing database or tracking systems.

Space Saving Design

  • Total height of less than 23mm — manual or automated storage capacity can be as much as doubled compared to higher volume tube options
  • Maximizing storage capacity allows improved utility of laboratory space and energy consumption

Innovative Cap Design

  • 200μl 2D internally-threaded universal cap features single-piece construction with molded-in-gasket that cannot be over-tightened manually or with automated capping tools
  • Thermoplastic elastomer gasket material is compatible for storage at room temperature down to the vapor phase of liquid nitrogen

Secure Tracking

  • Permanently-bonded, unique 2D barcode is laser-etched onto the base of every Thermo Scientific Matrix 2D Barcoded ScrewTop Storage Tube for secure identification and sample tracking
  • A Thermo Scientific 2D barcode reader instantly decodes each tube's 2D barcode scanning the tube into any application or database, or integrates via several export options
  • New barcoded rack allows tracking of the rack in addition to tubes
  • Laser-etched barcodes and human readable text on three sizes of rack, plus 2D code on rack bottom
  • Once sample data is stored, just scan the tube to access sample information and identify the contents

Stringent Quality Control

  • Every 2D barcoded storage tube is scanned to ensure readability
  • All barcodes are checked against our database of previously assigned codes to prevent duplicates
  • Every tube is leak tested to ensure sample integrity and security

Knowledgeable Support

  • Our team has a strong reputation for supporting both equipment and consumables
  • Consult with us for recommendations to optimize sample storage processes
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Screw Top Tube
200 μL
480
Tube barcode: Non-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction
Sterile
Sterile & PCR Ready
Matrix 200µL 2D Internally-threaded Universal Tubes, V bottom, w/ caps, Bulk, Sterile (1 bag of 480 tubes)
Polypropylene (Medical Grade), Polycarbonate, Polypropylene, TPV, Santoprene
Bulk, 480 Tubes
Matrix™
6762.8 fl oz.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.