missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Matrix - Green (Bulk Matrix from FastRNA Green)

Matrix - Green (Bulk Matrix from FastRNA Green) has 1.4 mm ceramic spheres that are ready for CE IVD labeling of clinical applications. For research use only.

Brand:  MP Biomedicals™ 116040801

1188.20 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


Product Code. 15719190

  • 1277.30 € / 1kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • 1.4 mm ceramic spheres
  • Ready for CE IVD labeling of clinical applications
  • For Research use only
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Zirconium Oxide Coated Beads
0.06 in.
FastPrep™ instruments or any bead beater.
Ceramic spheres
Off White
1.4 mm
Bulk Packaged
1 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers