Learn More
Thermo Scientific™ Matrix™ Pipette Tips
Choose from a comprehensive selection of these pipette tips designed together with pipetters to ensure a perfect balance between security of the seal and ease of application/ejection.
88.04 € - 689.04 €
Specifications
Filtered Non-filtered
Product Line Matrix
View More Specs
Products 14
Product Code Brand Volume (Metric) Sterility Format Price Quantity & Availability  
Product Code Brand Volume (Metric) Sterility Format Price Quantity & Availability  
10551715
View Documents
Thermo Scientific™
7320
300 μL Non-sterile Bulk This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10704444
View Documents
Thermo Scientific™
7321-11
300 μL Non-sterile Rack This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10191963
View Documents
Thermo Scientific™
7322
300 μL Sterile Rack This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10090754
View Documents
Thermo Scientific™
7610
10 μL Non-sterile Bulk This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10479743
View Documents
Thermo Scientific™
7611
10 μL Non-sterile Rack This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11752298
View Documents
Thermo Scientific™
7612-11
10 μL Sterile Rack This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11721208
View Documents
Thermo Scientific™
7620
20 μL Non-sterile Bulk This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11781198
View Documents
Thermo Scientific™
7621
20 μL Non-sterile Rack This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10454813
View Documents
Thermo Scientific™
8041-11
1250 μL Non-sterile Rack This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10334833
View Documents
Thermo Scientific™
8050
1250 μL Non-sterile Bulk This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10263723
View Documents
Thermo Scientific™
7152
250 μL Sterile Rack This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10354833
View Documents
Thermo Scientific™
7441
125 μL Non-sterile Rack This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10324833
View Documents
Thermo Scientific™
7442
125 μL Sterile Rack This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10099642
View Documents
Thermo Scientific™
8042
1250 μL Sterile Rack This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Choose from a comprehensive selection of pipette tips. Thermo Scientific™ Matrix Pipette Tips are supplied in innovative new Thermo Scientific™ Flo-Thru™ racks. Matrix pipetters and pipet tips are designed together to ensure that a perfect balance exists between security of the seal and ease of application/ejection. Manufactured under tight QC specifications, these standard tips rival special low-retention tips currently on the market after pipetting fluids such as dH2O, 50% Glycerol, Hybridized Buffer and 0.1% SDS.
Benefits:
  • Sealing rings on tips ensure confident, leak-free seal without banging tips on pipetter.
  • Because the sealing rings compress, these tips also offer a secure seal on most other pipetters on the market.
  • Highly polished molds and rigorous maintenance procedures ensure low liquid retention tips without special treatments or additives.
  • Stabilization rings in racks hold tips steady when seating on pipetter.
  • Venting features in rack ensure proper heat flow during autoclaving and reduce condensation on tips.
  • Finger notch on racks allows easy removal/insertion of refill trays.
Innovative Racks and Packaging:
  • Rigid rack design ensures solid, even tip seating; rack will not flex or bend when trying to apply tips.
  • Unique Thermo Scientific™ Flo-Thru™ rack individually supports tips to prevent them from wobbling when trying to seat them on the pipetter.
  • Hinge prevents sterile lid from coming in contact with lab bench.
  • Venting along the edge or around each tip improves heat flow during autoclaving; reduces condensation that can form on the tips.
  • Dual-position lid can be propped open to prevent vacuum deformation during autoclaving.
  • Sterile racks are sold in individual bags with quick-tear openings; each rack remains sterile until ready for use.
  • Each rack is labeled with product details including re-ordering information, description, volume range, and lot number.
  • Racks are constructed of clean recycled materials and are made as small as possible to reduce waste.
  • Racks are marked with appropriate symbols to support recycling initiatives.
Specifications
Specifications
Non-filtered
Matrix
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Matrix™ Pipette Tips