Learn More
Thermo Scientific™ Matrix™ Reagent Reservoirs
GREENER_CHOICE
Accommodate a variety of applications with these uniquely designed, innovative reagent reservoirs.
Brand:  Thermo Scientific™ 8093-11
112.00 EUR valid until 2025-05-31
Use promo code "25076" to get your promotional price.
 View more versions of this product
Product Code. 10756354
  • 212.00 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Accomodate a variety of applications with uniquely designed, innovative Thermo Scientific™ Matrix Reagent Reservoirs. These sterile reservoirs offer an extra-wide base to add rigidity and stability, helping to avoid spills. The 25mL divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizing recovery if using only a single channel and a small amount of reagent.
  • Trough within a trough maximizes amount of liquid accessible to pipet tips when using small amounts of reagent
  • Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
  • Graduations on inside wall enable quick measurement of remaining liquid
  • Sterile reservoirs are packaged in snap and tear bags, maintaining sterility while being easy to open
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25 mL
Sterile
10 Bags of 10
Polystyrene
Reagent Reservoir
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number21140949 Certificate TypeCertificate of Analysis Date11/11/2021 Catalog Number8093-11
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Matrix™ Reagent Reservoirs > Matrix™ Reagent Reservoirs; Volume: 25 mL; Material: Polystyrene; Sterility: Sterile; Packaging: 10 pcs /bag; Unit Size: 10 bags of 10 pcs (100 pcs)