missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Matrix™ ScrewTop Tube Caps, Cap Trays and Cap Inserts
GREENER_CHOICE

Simplify sample storage with these color-coded caps and inserts, available racked in trays or in bulk to suit a variety of applications.

Brand:  Thermo Scientific™ 4477BLU

 View more versions of this product

Product Code. 10290740

  • 394.00 € / Pack of 480
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Simplify sample storage with these color-coded caps and inserts. Thermo Scientific™ Matrix™ ScrewTop Tube caps and inserts are available racked in trays or in bulk to suit a variety of applications.

Superior Cap Design

  • Gasketed caps ensure critical sample protection over the storage lifetime
  • Soft gasket compresses to ensure a proper seal, preventing contamination from outside the tube
  • Cap material: Virgin class VI medical grade polypropylene with virgin class VI medical grade silicone O-ring
  • ScrewTop caps are compatible with storage down to -196ºC

Cap Options Simplify Sample Identification

  • Cap trays feature standard microplate format compatible with automated equipment
  • Caps are available in seven color options to enable sample identification
  • Color inserts snap into cap for a secure fit; inserts are removable
  • Inserts are available in six colors

All caps and inserts are supplied free from DNA, RNase, DNase, endotoxins and cytotoxins

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Croygenic Tube Closure
ScrewTop Tube Cap Tray
Blue
Matrix™
Sterile & PCR Ready
Autoclavable
Polypropylene, Silicone
5 Trays of 96 Caps
Sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.