missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Matrix™ SepraSeal and DuraSeal Capping Systems
GREENER_CHOICE

Accommodate a range of application and storage requirements. These flexible capping options allow automated piercing, visual identification and sterility preference.

115.00 € - 491.00 €


Products 29
Product Code Brand Product Line Description Color Price Quantity & Availability  
Product Code Brand Product Line Description Color Price Quantity & Availability  
11373772
GREENER_CHOICE
View Documents
Thermo Scientific™
4463BLU
Matrix™ SepraSeal Mats, Blue Blue
116.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
11731218
GREENER_CHOICE
View Documents
Thermo Scientific™
4463GRE
Matrix™ SepraSeal Mats, Green Green
119.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
11751218
GREENER_CHOICE
View Documents
Thermo Scientific™
4463GRY
Matrix™ SepraSeal Mats, Gray Gray
119.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
11741218
GREENER_CHOICE
View Documents
Thermo Scientific™
4463PUR
Matrix™ SepraSeal Mats, Purple Purple
119.00 €
Pack of 960
Estimated Shipment: 11-04-2025
to see stock.
Add to basket
 
11711218
GREENER_CHOICE
View Documents
Thermo Scientific™
4463RED
Matrix™ SepraSeal Mats, Red Red
119.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
11721218
GREENER_CHOICE
View Documents
Thermo Scientific™
4463YEL
Matrix™ SepraSeal Mats, Yellow Yellow
115.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
11855462
GREENER_CHOICE
View Documents
Thermo Scientific™
4464BLU
Matrix™ SepraSeal Mats, Sterile, Blue Blue
131.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
11865462
GREENER_CHOICE
View Documents
Thermo Scientific™
4464GRE
Matrix™ SepraSeal Mats, Sterile, Green Green
131.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
11875462
GREENER_CHOICE
View Documents
Thermo Scientific™
4464GRY
Matrix™ SepraSeal Mats, Sterile, Gray Gray
131.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
11895462
GREENER_CHOICE
View Documents
Thermo Scientific™
4464PUR
Matrix™ SepraSeal Mats, Sterile, Purple Purple
131.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
11805472
GREENER_CHOICE
View Documents
Thermo Scientific™
4464RED
Matrix™ SepraSeal Mats, Sterile, Red Red
131.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
12094148
GREENER_CHOICE
View Documents
Thermo Scientific™
4465BLU
Matrix™ SepraSeal Mats, Pre-Split, Blue Blue
136.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
12987365
GREENER_CHOICE
View Documents
Thermo Scientific™
4465GRE
Matrix™ SepraSeal Mats, Pre-Split, Green Green
136.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
12997365
GREENER_CHOICE
View Documents
Thermo Scientific™
4465GRY
Matrix™ SepraSeal Mats, Pre-Split, Gray Gray
136.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
12907375
GREENER_CHOICE
View Documents
Thermo Scientific™
4465PUR
Matrix™ SepraSeal Mats, Pre-Split, Purple Purple
136.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
12004158
GREENER_CHOICE
View Documents
Thermo Scientific™
4465RED
Matrix™ SepraSeal Mats, Pre-Split, Red Red
136.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
12927375
GREENER_CHOICE
View Documents
Thermo Scientific™
4465YEL
Matrix™ SepraSeal Mats, Pre-Split, Yellow Yellow
136.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
11912251
GREENER_CHOICE
View Documents
Thermo Scientific™
4466BLU
Matrix™ SepraSeal Mats, Pre-Split, Sterile, Blue Blue
143.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
12947375
GREENER_CHOICE
View Documents
Thermo Scientific™
4466GRE
Matrix™ SepraSeal Mats, Pre-Split, Sterile, Green Green
143.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
12957375
GREENER_CHOICE
View Documents
Thermo Scientific™
4466GRY
Matrix™ SepraSeal Mats, Pre-Split, Sterile, Gray Gray
143.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
12967375
GREENER_CHOICE
View Documents
Thermo Scientific™
4466PUR
Matrix™ SepraSeal Mats, Pre-Split, Sterile, Purple Purple
143.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
12977375
GREENER_CHOICE
View Documents
Thermo Scientific™
4466RED
Matrix™ SepraSeal Mats, Pre-Split, Sterile, Red Red
143.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
12987375
GREENER_CHOICE
View Documents
Thermo Scientific™
4466YEL
Matrix™ SepraSeal Mats, Pre-Split, Sterile, Yellow Yellow
143.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
10161293
GREENER_CHOICE
View Documents
Thermo Scientific™
4465
Matrix™ SepraSeal Natural
151.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
10019552
GREENER_CHOICE
View Documents
Thermo Scientific™
4490
Matrix™ DuraSeal
136.00 €
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
10385213
GREENER_CHOICE
View Documents
Thermo Scientific™
4463
Matrix™ SepraSeal Natural
119.00 €
Pack of 960
In Stock
to see stock.
Edge
Add to basket
 
11826103
GREENER_CHOICE
View Documents
Thermo Scientific™
4466
Matrix™ SepraSeal Pre-Split, Sterile Natural
143.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
10233393
GREENER_CHOICE
View Documents
Thermo Scientific™
4464
Matrix™ SepraSeal Natural
131.00 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
10203633
GREENER_CHOICE
View Documents
Thermo Scientific™
4469
Matrix™ SepraSeal SepraSeal Cap Removal Tool
491.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Accommodate a range of application and storage requirements. Thermo Scientific™ Matrix™ SepraSeal and DuraSeal tube sealing solutions complement the innovative line of Thermo Scientific Matrix 2D and non-2D coded storage tubes. Available in a range of formats to allow automated piercing, visual identification and sterility preference to meet your individual storage needs.

Sample Access Options
  • Designed for storage down to -20°C, Matrix SepraSeal mats are available in sterile, non-sterile, solid, or pre-split, and a range of colors for quick visual sample identification
  • Solid seals can be removed for sample access with the Thermo Scientific Matrix SepraSeal cap removal tool, or can be pierced once
  • For applications involving repetitive sample access, a pre-split Matrix SepraSeal can be pierced by automated liquid handling equipment, requiring very little pressure, while maintaining seal integrity with self-sealing closure
  • For applications requiring storage down to -80°C, permanently installed DuraSeals are an ultra robust SepraSeal option—can also be used for repetitive piercing procedures
  • The Matrix DuraSeals Pre-Split V-Shaped design reliably and accurately guides automated liquid handling needles into the same location over multiple piercings—self-sealing design maintains sample integrity and prevents loss during storage

Application Flexibility

  • Elastomer ensures sealing performance will not degrade, even after multiple cap piercings
  • The Matrix SepraSeal tube sealing solution offers the flexibility of individual tube capping while also providing the ability to apply 96 caps at once
  • Supplied in mats of 96 caps each, sealing is secure and consistent every time when applied with the Thermo Scientific SuperSealer
  • After the SuperSealer applies the mat, simply pull up the backing sheet to reveal 96 individually sealed tubes—for applications where multiple piercings are required
  • The Matrix DuraSeal capping system is available in bulk or pre-installed in a wide array of our storage tubes
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.