Learn More
Thermo Scientific™ Matrix™ WellMate™ Disposable Tubing Cartridges
GREENER_CHOICE
Eliminate the need for service or calibration with disposable tubing cartridges. These cartridges dispense everything from sensitive cellular materials to high vapor pressure fluids.
Brand:  Thermo Scientific™ 201-30002
1239.00 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10117633
  • 2100.00 € / Pack of 5
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Eliminate the need for service or calibration with Thermo Scientific™ Matrix™ WellMate™ Disposable Tubing Cartridges. These cartridges dispense everything from sensitive cellular materials to high vapor pressure fluids. They are presterilized and can be easily and quickly replaced.
Features:
  • Disposable, irradiated, precalibrated tubing
  • Standard-bore nozzle: for volumes above 20 μL, and 6 to 96-well plate filling
  • Small-bore nozzle: for lower volume (1.0 μL and above) applications
  • Stainless steel, PTFE-coated: for use with viscous fluids, 6 to 384-well dispensing, and low-profile plates
Includes:
5 Cartridges per case
Compatible with:
WellMate base unit
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Disposable Tubing Cartridge
PTFE
5/Cs.
Tubing Cartridges; Small Bore
>1 μL