Learn More
Brady™ Matte Polyester RFID Chemical Resistant Slide Labels
UHF RFID labels for use on medical slides.
Brand:  Brady™ 309083
Product Code. 17445993
  • 728.00 € / 1 roll
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use in pharmaceutical industry and healthcare
  • Designed for use on medical slides in the laboratory environment
  • Polyester tag face material with chip bonded to aluminum antenna using anisotropic conductive film adhesive
  • Suitable for thermal transfer printing, including RFID encoding
  • Labels are outside wound and 100% inspected, 500 good RFID labels per roll with bad tags marked
Specifications
Specifications
Not Autoclavable
Permanent Acrylic
Matte
Thermal Transfer
RFID Labeling, Healthcare and Hospital Labeling, Laboratory Labeling
1
-80°C to +100°C
20 mm
16 mm
THT-UHF-481-24x16
White
Polyester
Non-sterile
Sato CL4NX Plus Industrial Label Printer, Zebra™ ZT411 Industrial Label Printer
500 Labels
0.29 mm
24 mm