Learn More
Thermo Scientific™ MaxiMix™ I Vortex Mixer
Mix quickly and evenly in continuous operation or touch-on mode with this mixer. Excellent for single and multiple tubes and small flasks with closed top.
96.50 €
Specifications
Speed Range | 0 to 3000 rpm |
---|---|
Product Type | Mixer |
Frequency | 50/60 Hz |
Material | Cast Aluminum Base, Foam Platform |
Dimensions (L x W x H) | 5.5 x 5.5 x 4.2 in. (13.9 x 13.9 x 10.6 cm) |
Product Code | Brand | Voltage | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Voltage | Price | Quantity & Availability | ||||||
13453726
|
Thermo Scientific™
M16710-33Q |
230 V | N/A | N/A | N/A | |||||
Description
Mix quickly and evenly in continuous operation or touch-on mode with the Thermo Scientific™ MaxiMix™ I Vortex Mixer. This mixer is ideal for enzymatic and RIA assays, general test tube mixing, biochemical assays, viral dilutions preparations, precipitated assays, cell suspension vortexing and tissue sample mixing. This mixer is excellent for single and multiple tubes, as well as small flasks with closed tops.
Features:- Simultaneously mixes up to four test tubes
- Push-button top-mounted ON/OFF switch
- Vary vortex mixing by simply changing pressure of tube against foam rubber top
- Durable, white plastic case resists acids and alkalis
- Cast-aluminum base with suction-cup rubber feet provides maximum stability
- Compact design with 4 in. diameter (10.2cm) foam rubber top
Includes:
- One extra foam top (12-814-1)
- Three-wire power cord and plug
Recommended for:
- Enzymatic and RIA assays
- General test tube mixing
- Biochemical and precipitated assays
- Vortexing cell suspensions
- Mixing tissue samples
- Viral dilution preparations
Warranty: 90 days
Specifications
0 to 3000 rpm | |
50/60 Hz | |
5.5 x 5.5 x 4.2 in. (13.9 x 13.9 x 10.6 cm) | |
10.6 cm | |
13.9 cm | |
13.9 cm | |
1.5 kg | |
MaxiMix I Vortex Mixer | |
90 Days (Labor), 1 Year (Parts and Labor) |
Mixer | |
Cast Aluminum Base, Foam Platform | |
4.2 in. | |
5.5 in. | |
5.5 in. | |
3.5 lb. | |
Push Button On/OFF Switch | |
Touch, Continuous |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.