Learn More
Thermo Scientific™ Maxima Hot Start Taq DNA Polymerase
Enhance the specificity, sensitivity, and yield of DNA amplification with this hot start DNA polymerase.
57.25 € - 878.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10394380
|
Thermo Scientific™
EP0602 |
500U | N/A | ||||||
10763721
|
Thermo Scientific™
EP0603 |
1000U | N/A | ||||||
10140740
|
Thermo Scientific™
EP0601 |
N/A | |||||||
11886833
|
Thermo Scientific™
K1051 |
2 x 1.25mL | N/A | ||||||
11883923
|
Thermo Scientific™
K1052 |
10 x 1.25mL | N/A | ||||||
Description
Maxima Hot Start Taq DNA Polymerase is a chemically modified recombinant Taq DNA polymerase. The enzyme is inactive at room temperature, avoiding extension of non-specifically annealed primers or primer dimers and providing higher specificity of DNA amplification. The functional activity of the enzyme is restored during a short 4 minute incubation at 95°C. The activated enzyme maintains the same functionality as Taq DNA polymerase. Maxima Hot Start Taq DNA Polymerase is also available in a master mix format.
Highlights
- Four minute activation time
- High PCR specificity and sensitivity
- Room temperature PCR set up